Learn More
Chemglass Life Sciences Quartz Wool, Coarse, Fiber Size 8-15 Microns
Supplier: Chemglass Life Sciences CGQ069002
Quartz Wool, Coarse, Fiber Size 8-15 Microns. Useful as high temperature packing in distillation columns or as a bed support for high purity liquid chromatography. Coarse porosity. SOLD IN 1/2 LB. PACKAGES.