Learn More
Starna Cells Inc QUARTZ CUVETTE
Supplier: Starna Cells Inc 18BQ10
Micro Spectrophotometer Cells Self Masking Black Walls Path Length: 10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 45mm Interior Width: 2mm Interior Length: 10mm Nominal Volume: 0.700mL Number of Polished Windows: 2