Learn More
Anton Paar QUARTZ CONTROL PLATE, +50°Z/+17°OR for MCP 100
Supplier: Anton Paar 143960
Quartz control plate with +50°Z/+17°Optical Rotation used for checking and adjusting the polarimeter scale on the MCP 100.Suitable for use with Anton Paar polarimeter MCP 100 item NC1818306