Learn More
Charles Supper Co Inc Thin-Walled Capillary Tubes for X-Ray Diffraction - Quartz (QZ) 1.5mm, 25/pack
Supplier: Charles Supper Co Inc QUARTZ 15QZ
Wall Thickness – 0.01mm. Approximate Overall Length 80mm. Drawn from Clear, Fresh, Properly Cleaned Glass. Funnel at one end and Sealed at the other end. Supplied in non confusing, color coded glass packages containing 25 tubes. Individual tubes are easily removed from glass package without fumbling and searching through the container.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.