Learn More
Quantimetrix QuanTscopics, Urinalysis Microscopics Control, Level 1
Supplier: Quantimetrix 148141
•Cost-effective•Liquid, made with human urine•2-year shelf life & open vial stability from date of manufacture when stored at 2ºC–8ºCFormulated with real human RBCs and WBCs, QuanTscopics makes for a cost-effective way to monitor the performance of urinalysis sediment for both manual and automated microscopy methods. We include qualitative and quantitative values for standardized urinalysis systems and analyzers. The control features easy-to-use squeeze bottles for simple dispensing.Thoughtfully redesigned packaging now includes barcodes on the labels for lot numbers and expiration dates and we’ve made it easier to identify each level.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.