Learn More
CTL Scientific Supply Corp QUANTOFIX SULFATE
Supplier: CTL Scientific Supply Corp 91329
QUANTOFIX SULFATE 100 strips/pack Test strips used for the semi-quantitative determination of sulphate in solutions. Gradation of scale: <200 - > 400 - > 800 - > 1200 - > 1600 ppm SO42-