Learn More
CTL Scientific Supply Corp QUANTOFIX LUBRICHECK
Supplier: CTL Scientific Supply Corp 91336
Test sticks for testing concentration of cooling lubricants can quickly be determined. Cooling lubricant can easily be checked. Ensures optimal cooling and lubrication and at the same time optimal quality of workpiece. 0, 15, 50, 75, 130, 200 mmol/l