Learn More
Sartorius Quantitative Grade 390 Filter Papers
These filter papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration. They are made out of 100 % cotton linters with an α-cellulose content of > 98 % and are acid-washed to make the papers ashless and achieve high purity.
$13.90 - $172.20
Catalog Number | Mfr. No. | Format | Filtration Speed | Particle Retention | Diameter (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Filtration Speed | Particle Retention | Diameter (Metric) | Price | Quantity & Availability | ||
14-555-221
|
Sartorius
FT3103110 |
Disc | 100 sec. | 3 to 5 μm | 110 mm |
Pack of 100 for $42.95
|
|
||
14-555-220
|
Sartorius
FT3103125 |
Disc | 100 sec. | 3 to 5 μm | 125 mm |
Pack of 100 for $59.70
|
|
||
14-555-218
|
Sartorius
FT3103185 |
Disc | 100 sec. | 3 to 5 μm | 185 mm |
Pack of 100 for $104.80
|
|
||
14-555-219
|
Sartorius
FT3103150 |
Disc | 100 sec. | 3 to 5 μm | 150 mm |
Pack of 100 for $96.50
|
|
||
14-555-014
|
Sartorius
FT4103150 |
Folded | 100 sec. | 3 to 5 μm | 125 mm |
Pack of 100 for $91.00
|
|
||
14-555-224
|
Sartorius
FT3103055 |
Disc | 100 sec. | 3 to 5 μm | 55 mm |
Pack of 100 for $13.90
|
|
||
14-555-222
|
Sartorius
FT3103090 |
Disc | 100 sec. | 3 to 5 μm | 90 mm |
Pack of 100 for $32.90
|
|
||
14-555-223
|
Sartorius
FT3103070 |
Disc | 100 sec. | 3 to 5 μm | 70 mm |
Pack of 100 for $17.63
|
|
||
14-555-015
|
Sartorius
FT4103125 |
Folded | 250 sec. | 1 to 2 μm | 125 mm |
Pack of 100 for $80.90
|
|
||
14-555-217
|
Sartorius
FT3103240 |
Disc | 100 sec. | 3 to 5 μm | 240 mm |
Pack of 100 for $172.20
|
|
||
Description
- Ash-free (ash content ≤ 0.01 % according to DIN 540370)
- Wet-strengthened
- Color-coded box for easy selection
Specifications
Green | |
390 | |
100/Pk. |