Learn More
Invitrogen™ QuantiGene™ Sample Processing Kit, tissues
The QuantiGene Sample Processing Kit for tissues contains reagents and instructions for the preparation of tissue homogenates from fresh or frozen animal or plant tissues for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
$138.00 - $674.00
Specifications
Format | Kit |
---|---|
Sample Type | Tissue |
Product Line | QuantiGene™ |
Product Type | Sample Processing Kit |
Shipping Condition | Blue Ice |
Description
The QuantiGene Sample Processing Kit for tissues contains reagents and instructions for the preparation of tissue homogenates from fresh or frozen animal or plant tissues for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
- Easy to follow instructions
- Preserves the integrity of the RNA/DNA without degradation
- Choose the method that matches your tissue sample
What you get
The QuantiGene Sample Processing Kit for tissues contains our proprietary homogenizing solution, proteinase K, and detailed instructions for preparing homogenized tissue from a variety of tissue sources.
Specifications
Kit | |
QuantiGene™ | |
Blue Ice |
Tissue | |
Sample Processing Kit |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.