Learn More
Electron Microscopy Sciences QUANTIFOIL R 1.2/1.3, Gold, 200 mesh, 100/pack
Supplier: Electron Microscopy Sciences Q2100AR1.3
A foil with ~1.2 µm circular holes and a spacing of ~1.3 µm between the holes. This type is used at magnifications around 50,000x.