Learn More
Bioassay Systems QuantiChrom™ Peroxide Assay Kit. For quantitative determination of peroxide and evaluation of drug effects on peroxide metabolism.
SDP
Supplier:  Bioassay Systems DIOX250
Encompass_Preferred
For quantitative determination of peroxide and evaluation of drug effects on peroxide metabolism. Key Features: Sensitive and accurate. Enhanced color intensity using sorbitol. Detection range 0.2 µM (7 ng/mL) to 30 µM (1,020 ng/mL) H2O2 in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single detection reagent and incubation for 30 min. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Method: OD585nm. Samples: Serum, citrate-plasma, urine, cell lysate, culture media etc. Species: All. Procedure: Assay takes 30 min. Kit size: 250 tests. Detection limit: 0.2 µM.
Catalog No. 50-489-222