Learn More
CTL Scientific Supply Corp QUANT. TOTAL IRON 1000,100 STR
SDP
Supplier:  CTL Scientific Supply Corp 91330
Encompass_Preferred
QUANTOFIX TOTAL IRON 1000 are test sticks and reagents for the semi-quantitative determination of iron (Fe 2+ /Fe 3+ ) in solutions. Gradation of scale: 0-5-20-50-100-250-500-1000 mg/l Fe 2+/3+ white to dark red. Contains 100 test strips
Catalog No. 50-117-3318
Explore available promotions