Learn More
CTL Scientific Supply Corp QUANT. NITRATE 100,100 TESTS
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 91351
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
QUANTOFIX NITRATE 100 are test sticks for the semi-quantitative determination of nitrite (NO2) in solutions Gradation of scale: 0-5- 10-25-50-75-100 mg/l. white to red-violet. Contains 100 test strips