Learn More
CTL Scientific Supply Corp QUANT. CHLORINE SENS.,100 STR
SDP
Supplier:  CTL Scientific Supply Corp 91339
Encompass_Preferred
QUANTOFIX CHLORINE SENSITIVE are test sticks for the semi-quantitative determination of total chlorine. Gradation 0 - 0.1 - 0.5 -1 - 3 - 10 mg/l. Contains 100 test strips
Catalog No. 50-117-3319