Learn More
MilliporeSigma™ Qualification Workbook
Provides instructions on performing system qual and completing qual forms
Supplier: MilliporeSigma™ VMQSX0FTB
Includes: IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures
Description
- For use with Milli-RO, Milli-RX, Milli-Q, RiOs, SDS, Elix, Direct-Q systems
Specifications
Milli-Q Biocel System (Y) | |
Qualification Workbook | |
Provides instructions on performing system qual and completing qual forms | |
IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures |