Learn More
Mopec™ Qlicksmart BladeFlask EVO Scalpel Blade Remover
The Qlicksmart BladeFlask EVO can accommodate a broad range of scalpel handles and blades, including large autopsy scalpels, thick and round handles, and even hexagonal shaped versions. This gives you greater freedom in using any scalpel you want.
Supplier: Mopec™ AJ160
Description
- Simple to use, even with just one hand
- Embrace safety with this new standard in scalpel blade removals
- Blade Capacity: 100 Blades
- Compatible with all commercially available scalpel blades, including size PM60A, PM60B, and PM70
- Compatible with all commercially available scalpel handles, including PM6 and PM8
- Easy mounting with optional mounting bracket
Specifications
Scalpel Blade Remover | |
Reusable | |
OSHA Compliant |
Universal | |
Sterile | |
No |