Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QIAGEN QIAcard FTA Mini
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
$277.00 - $1103.00
Specifications
Description | Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Mini Cards. |
---|---|
Sample Type | Blood |
For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology |
No. of Samples | 2 |
Product Type | FTA Mini Card |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
09-800-994
|
QIAGEN
WB120355 |
25 Pk. |
Pack for $277.00
|
|
|||||
09-923-342
|
QIAGEN
WB120055 |
100 Pk. |
Pack of 100 for $1,103.00
|
![]() |
|||||
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
- QIAcard FTA Mini format with 2 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
- FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Mini Cards. | |
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology | |
FTA Mini Card |
Blood | |
2 | |
125 μL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title