Learn More
World Precision Instrument QFT1 Filter Holder for Glass Fiber Filters
SDP
Supplier:  World Precision Instrument 89575
Encompass_Preferred
Designed for field use
WPI’s filter holder for particulate absorption measurements is specially designed for field use. It is rugged and portable. It performs as well as a laboratory based spectrophotometer. It can be directly connected to WPI’s line of fiber optic spectrometers and light sources. Instead of collecting your samples, transporting them to a laboratory, and accepting the loss of spectral information associated with it (Sosik, 1999), particulate absorption can now be measured on site.
Catalog No. 50-252-1218