Learn More
Thermo Scientific™ AcroMetrix NAT Blood Screening Controls
Assess performance of NAT assays that test human plasma from donations of whole blood and blood components with these controls, which are for In Vitro Diagnostic Use.
$351.00
Specifications
Sample Type | Plasma |
---|---|
Type | Control |
Packaging | 10 x 1.4mL |
Quantity | 10 x 1.4 mL |
Catalog Number | Mfr. No. | Description | Detectable Analytes | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Detectable Analytes | Packaging | Price | Quantity & Availability | |||
23-279-180
|
Thermo Scientific™
960500 |
AcroMetrix WNVRNA Control | West Nile Virus (WNV) | 10 x 1.4mL |
Pack of 10 for $351.00
|
|
|||
Description
Estimate the precision and reproducibility of NAT assays that are designed to detect viral nucleic acids in human plasma from donations of whole blood and blood components for transfusion with the Thermo Scientific™ AcroMetrix NAT Blood Screening Controls. These controls mimic naturally occurring plasma specimens and can help detect potential systematic deviations of tests for the qualitative determination of viral nucleic acids. The controls are for In Vitro Diagnostic Use.
Specifications
Plasma | |
Control | |
10 x 1.4mL | |
10 x 1.4 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.