Learn More
QBC Diagnostics AccuTubes
AccuTubes can be used for capillary samples from a finger or heel stick, as well as venous blood samples.
Supplier: QBC Diagnostics 423406
Description
- They are packaged with a ventilated stopper inserted into the tube and a soft grip float holder for easy insertion of the plastic float.
- For use with the Autoread or Autoread Plus.
Specifications
Blood Collection Tube | |
Autoread Plus Hematology System | |
Ventilated stopper inserted into the tube, and a soft grip float holder for easy insertion of the plastic float | |
CLIA EQC Option 2 |
70 μL | |
100 Pk. | |
Used for capillary samples from a finger or heel stick, as well as venous blood samples | |
70 μL |