Learn More
Bioassay Systems QuantiQuik™ Urea (BUN) Quick Test Strips. Direct determination of urea concentrations in food and beverage samples as well as biological samples.
Supplier: Bioassay Systems QQUREA10
For rapid, direct determination of urea concentrations in food and beverage samples as well as biological samples. Key Features: Fast and sensitive. Use of 20 µL sample. Semi-quantitative measurement between 0-1500 mg/L (undiluted) urea. Convenient. No expensive plate or cuvette readers needed. Sample trea™ent and assay can be performed in under 15 minutes. Method: Visual. Samples: Serum, plasma, urine, etc. Species: All. Procedure: Assay takes approximately 15 min. Kit size: 10 tests.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.