Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Q-Bio Taq DNA Polymerase
Q-BioTaq™ DNA Polymerase is a thermostable DNA polymerase which can be used effectively for PCR and DNA sequencing by the chain termination method.
Supplier: MP Biomedicals™ 11EPQBT100
Description
- For multiplex PCR and RAPDs
- Recombinant, truncated form of Thermus aquaticus
- Highly thermostable (98°C)
- No 5'-3' exonuclease activity
- Enzyme cannot degrade the 5' end of primers
- This enzyme is encoded by a modified form of the Thermus aquaticus DNA polymerase gene with an N-terminal deletion.
- The properties of QBioTaq™ DNA Polymerase include high thermostability and absence of 5'-3' exonuclease activity.
- This polymerase is particularly useful in applications where multiple amplifications are required.
- The enzyme is highly purified and free of RNases and endo- and exonucleases.
Specifications
Q-Bio Taq Pol Bulk | |
1000 u |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction