Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyridine (Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Can
Glass Bottle
Specifications
pH | 8.5 |
Melting Point | -42°C |
Color | Colorless |
Density | 0.978g/cm³ |
Boiling Point | 115°C |
Quantity | 4 L |
Identification | Pass Test |
Packaging | Aluminum Can |
Physical Form | Liquid |
Assay Percent Range | ≥99 % |
Show More |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. NFPA Health:2 Flammability:3 Instability:0 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction