Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrantel Pamoate, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog Number 18605965
Click to view available options
Quantity:
25 g
100 g
1 kg
20 kg
Packaging:
Amber Glass Bottle
Fiber Drum
22204-24-6
C34H30N2O6S
594.68
AQXXZDYPVDOQEE-MXDQRGINSA-N
1-methyl-2-[(1E)-2-(thiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine; 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
CN1CCCN=C1\C=C\C1=CC=CS1.OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1
22204-24-6
C34H30N2O6S
594.68
AQXXZDYPVDOQEE-MXDQRGINSA-N
1-methyl-2-[(1E)-2-(thiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine; 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
CN1CCCN=C1\C=C\C1=CC=CS1.OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1

P1599, 22204-24-6, C11H14N2S•C23H16O6

Spectrum™ Chemical Pyrantel Pamoate, USP is an anthelmintic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Ignition Residue 0.005
Loss on Drying 0.02
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 97 to 103%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit