Learn More
Thermo Scientific™ PVDF Transfer Membranes, 0.45 μm
Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for western, Southern, and northern blotting methods.
$261.50 - $674.00
Specifications
Material | PVDF |
---|---|
Pore Size | 0.45 μm |
Content And Storage | Store membranes flat at ambient temperature and away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure. |
Shipping Condition | Room Temperature |
Catalog Number | Mfr. No. | Dimensions (LxW) | Format | Pore Size | Sufficient For | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (LxW) | Format | Pore Size | Sufficient For | Price | Quantity & Availability | ||
PI88585
|
Thermo Scientific™
88585 |
10 x 10 cm | Sheet | 0.45 μm | 10 Mini-gel Blots |
Each for $261.50
|
|
||
PI88518
|
Thermo Scientific™
88518 |
3.75 m x 26.5 cm | Roll | 0.45 μm | ≥100 Mini-gel Blots (When cut to 8.8 x 10 cm); ≥80 Midi-gel Blots (When cut to 8.5 x 13.25 cm) |
Each for $674.00
|
|
||
Description
Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for western, Southern, and northern blotting methods. The 0.45-μm pore size make these membranes well-suited for a wide range of western blotting conditions and ideal for transfer of proteins >10 kDa. For best results, preactivate the membrane with 100% alcohol.
Features include:
• Re-probe characteristics—high mechanical strength makes PVDF an excellent membrane for stripping and re-probing
• Durability—compatible with most organic solvents, acids, and mild bases
• Compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
• High sensitivity—exceptional binding capacity makes PVDF the ultimate choice for low abundance proteins
Specifications
PVDF | |
0.45 μm | |
Store membranes flat at ambient temperature and away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure. | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.