Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ PVC-Coated Wire Rack for 60mL BOD Bottles
This rugged wire rack holds up to (20) 60mL bottles for drying or storage.
Supplier: DWK Life Sciences W227729
Description
- Manufactured from rugged PVC coated wire
- Coating is corrosion resistant
- Racks interlock to connect stacked racks for safe transport
- Made in the United States
Specifications
Bottle Rack | |
Bottles (20 x 60 mL) | |
Yes - Interlocking | |
1 Each |
Corrosion-resistant rack | |
PVC Coated Wire | |
BOD bottles |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction