Learn More
United Scientific Supplies Push Pull Scale
Push Pull Scale for measuring weights, masses and forces.
Supplier: United Scientific Supplies PS2000
Description
- Spring scale can be used to measure forces using push or pull functions
- To weigh suspended object from large hook
- Includes zero adjustment screw and metal suspension ring
- Available in six color-coded sizes
Specifications
Push Pull Scale | |
Red | |
For measuring weights, masses and forces | |
2000 g |
20 N | |
0.4 N | |
40 g |