Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Purifier™ Cell Logic™+ Class II B2 Biosafety Cabinets with Scope-Ready, Temp-Zone and Base Stand
Microscope-ready B2 biohazard cabinets with advanced ergonomic design excel in user comfort and functionality
$30401.00 - $34239.00
Specifications
Chamber Material | Stainless Steel |
---|---|
Product Type | Type B2 Biosafety Cabinet |
Model | Purifier Cell Logic+ |
Noise Level | <63 dBA |
Receptacles | 2 |
Catalog Number | Mfr. No. | Width (English) Interior | Outlets | Plug Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) Interior | Outlets | Plug Type | Price | Quantity & Availability | |||
34-348-1101
|
Labconco™
343481101 |
4 ft. | 4 | North America (115 V, 15 A) |
Each for $31,360.50
|
|
|||
34-348-1111
|
Labconco™
343481111 |
4 ft. | 2 | North America (230 V) |
Each for $30,401.00
|
|
|||
34-348-1121
|
Labconco™
343481121 |
4 ft. | 4 | Japan (100 V, 15 A) |
Each for $30,401.00
|
|
|||
34-348-1131
|
Labconco™
343481131 |
4 ft. | 2 | Schuko (230 V) |
Each for $30,401.00
|
|
|||
34-348-1141
|
Labconco™
343481141 |
4 ft. | 2 | China, Australia (230 V) |
Each for $30,401.00
|
|
|||
34-348-1151
|
Labconco™
343481151 |
4 ft. | 2 | UK (230 V) |
Each for $30,401.00
|
|
|||
34-348-1171
|
Labconco™
343481171 |
4 ft. | 2 | India (230 V) |
Each for $30,401.00
|
|
|||
34-368-1101
|
Labconco™
343681101 |
6 ft. | 4 | North America (115 V, 15 A) |
Each for $34,239.00
|
|
|||
34-368-1111
|
Labconco™
343681111 |
6 ft. | 2 | North America (230 V) |
Each for $34,239.00
|
|
|||
34-368-1121
|
Labconco™
343681121 |
6 ft. | 4 | Japan (100 V, 15 A) |
Each for $34,239.00
|
|
|||
34-368-1131
|
Labconco™
343681131 |
6 ft. | 2 | Schuko (230 V) |
Each for $34,239.00
|
|
|||
34-368-1141
|
Labconco™
343681141 |
6 ft. | 2 | China, Australia (230 V) |
Each for $34,239.00
|
|
|||
34-368-1151
|
Labconco™
343681151 |
6 ft. | 2 | UK (230 V) |
Each for $34,239.00
|
|
|||
34-368-1171
|
Labconco™
343681171 |
6 ft. | 2 | India (230 V) |
Each for $34,239.00
|
|
|||
Description
All models feature:
- Electronically Commutated Motor (ECM)
- Constant Airflow Profile (CAP) airflow monitoring system
- Intrinsically-safe negative pressure design
- Air-Wave Entry System
- Contain-Air Negative Pressure Channel
- Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
- Interior-mounted, line-of-sight, color LCD with MyLogic OS displays filter life, status messages for alarm conditions and alerts. 8 languages available
- Bright, 90 to 150 footcandle, glare-free LED lighting located outside the contaminated work area
- Fully-closing, clear 1/4 in. tempered safety glass sash
- Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
- Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
- Smart-Start™ System user-programmable start up and shut down
- Leak-tight stainless steel interior and powder-coated steel exterior
- Removable towel catch located under work surface
- Code-activated electronic security lock
- 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
- 10° Angled sash with counterbalanced, easy lift mechanism
- ADA-Compliant touchpad control on right side post
- Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
- Scope-Ready™ Package consisting of 7 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar. Stand-Still™ Isolation Platform is required and sold separately
- Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. (27 cm) wide x 12.0 in. (30 cm) deep (7.0 x 12 in. on 3 ft. models) located on the left side of the work surface. Circulator is required (not included). Circulator requirements: minimum flow rate of 0.4 to 0.5 GPM (1.5 to 1.9 LPM), typical volume of 6 liters, and temperature of 0 to 50° C (chiller/heater)
- Noise level <63 dBA
- Heat load 990 BTU/hour (4 ft. models)
- Nominal inflow velocity of 105 fpm (0.5 m/sec)
- Nominal downflow velocity of 55 fpm (0.3 m/sec)
- 10 ft. (3 m) power cord with plug
- Five year warranty
- NSF International compliant
- Optional NSF-approved non-welded telescoping base stand included on some models
- Optional Accessory Package (ADA-compliant service fixture(s) with ball-type valve(s) and NSF-Approved Vacu-Pass™ Cord and Cable Portal) included on some models
- All models require supporting base if non-welded stand option is not selected
Specifications
Stainless Steel | |
Purifier Cell Logic+ | |
2 | |
20.3 cm | |
LED | |
8 in. | |
31.2 in. |
Type B2 Biosafety Cabinet | |
<63 dBA | |
100.1 to 106.1 in. | |
Cabinet with Accessory, Temp-zone and Scope-ready Packages, Base Stand | |
254.2 to 269.2 cm | |
5 Years | |
79.248 cm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title