Learn More
Uline Honeywell Pure Flow 1000 Saline Cartridge,Set of two 3 12 Gal Saline Cartridges
Supplier:  Uline S11507
Encompass
Saline cartridges for use with Pure Flow 1000 Eye Wash Station, 24-month cartridge life 4 times longer than Porta Stream II, Meets ANSI Z3581-2014
Catalog No. NC2312700