Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gentex™ PureFlo ESM+ PF60 PAPR Respirator System, Black Hard Hat
Self-contained PAPRs are ideal for medium to heavy-duty applications. Innovative all-in-one design provides head, eye, face, and respiratory protection against hazardous airborne contaminants, sparks, flying debris, chemical and molten metal splashes.
Supplier: Gentex™ PR05801B0C1
This item is not returnable.
View return policy
Description
- Includes ANSI-approved hard hat, HEPA filter, battery pack, visor assembly, and cape
- Charger sold separately
- Electronic Safety Management System (ESM+) includes LED status indicator and audible alarm for low flow, low battery, and filter change
- Adjustable flame retardant neck cape
- No cables, hoses, or belt-mounted blower
- Unique continuous airflow system with airflow rate of 8.1 cfm
- Wide vision ANSI approved polycarbonate face shield
- Quick-release headband for easy adjustment and donning
- HEPA (HE) Filter (other filters available)
- Corrosion-resistant gold plated battery
- Choose from internal and external on/off switch or internal switch only
Specifications
Head Protection: ANSI Z89.1 Type 1 Class G; Face Shield/Eye Protection ANSI Z87.1+: 2015 Certified; SEI Certified and Listed | |
7 hours with two batteries | |
PARTICULATE | |
PAPR Hard Hat System | |
HEPA (HE) or HEPA/HF/HC *Nuisance OV | |
Black Hard Hat, Black Stedprene FR Neck Cape, HE/HF/HC Filter, APF 1000, Internal Switch | |
Under 70 dB | |
2.5 hr. |
1000 | |
5 Cell Nickel Metal Hydride | |
NIOSH Approved | |
One Size | |
8.1 CFM or 230 L/min. | |
PR05801-B-0C1 | |
1 Ea. | |
One Size |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction