Learn More
StatLab™ PureFix™ Fixative, 10% Non-buffered Formalin
A 10% non-buffered formalin preferred by particular labs.
Supplier: StatLab™ 31805
Alert:
Harmful if swallowed, if in contact with skin, or if inhaled.
Description
- Will not produce salt precipitates that may obstruct tissue processor lines
Specifications
Cube | |
5 gal. | |
Tissue Fixation |