Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUCHI Pure Cartridge Holders
For use with Pure Chromatography Systems
$999.00 - $2283.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
05-405-472
|
BUCHI
11065940 |
Cartridges (4 to 330 g) |
Each for $999.00
|
|
|||||
05-405-544
|
BUCHI
11070532 |
Cartridges (>300 g) |
Each for $2,283.00
|
|
|||||
Description
- For use with BUCHI Pure Chromatography Systems
- Available in two sizes to accomodate a wide variety of cartridges
Specifications
Cartridge Holder |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title