Learn More
Air Science Usa Air Science Purair™ Horizontal Laminar Flow Cabinet Clean Bench, 48" / 1200mm Nominal Width, 115V 60Hz, Extra Tall Height Option
Supplier: Air Science Usa HLF48XT
Purair LF Series Horizontal Laminar Flow Cabinet / Clean Bench (HLF-48-XT), 48" / 1200mm Nominal Width, Extra Tall Internal Height (30"), 115V 60Hz (other voltages available). PurairHorizontal Laminar Flow Hoods are a series of high efficiency products designed to protect equipment and other contents of the work zone from particulates, for applications sensitive to such contamination. It is ideally suited for use with non-hazardous contaminants and when flexible access to the equipment in the work zone is desired. At the heart of the Purair Laminar Flow Hood product line is the Air Science Multiplex ULPA Filtration Technology that creates a clean work environment over a wide range of applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.