Learn More
Hach Company Pump, Vacuum, Hand-operated
Supplier: Hach Company 1428300
This unit is ideal for field use and is a real convenience in the lab. Pumping rate is 15 cc (0.50 oz) per stroke. Entire unit is corrosion-resistant, sealed, and self-lubricating. Nozzle fits standard 6 mm (1/4") I.D. tubing. Supplied with 61 cm (2') of clear plastic tubing. Choose with or without vacuum gauge.