Learn More
Cole-Parmer Air Cadet Vacuum/Pressure Pump, Diaphragm, Single Head, 0.7 cfm, 12 VDC
Supplier: Cole-Parmer 0753225
This all-plastic, convoluted diaphragm design handles many vacuum or pressure applications, while reducing stress, wear and heat build-up. The pumps are available with standard 115 or 230 VAC motor; or portable 12 VDC motor. Standard AC motors are service-free, brushless type with sealed ball bearings, and thermal overload protection.