Learn More
Adam Equipment Adam Equipment PTM Drum and Wheelchair Scale PTM 500 1100lb / 500kg capacity x 0.2lb / 0.1kg readability
Supplier:  Adam Equipment PTM 500 AE403A
Encompass_Preferred
"Adam Equipments PTM wheelchair scale features a low-profile base and easy-access ramps on both sides, offering smooth transitions from the floor to the platform and is suitable for use in warehouses or medical facilities. PTM drum and wheelchair scales are ideal for checking the weight of wheelchair occupants, barrels and drums, or loaded dollies. Its diamond plate surface provides good grip and a solid, steady weighing area. Handles and wheels enable easy accessibility in a wide range of environments from doctors surgeries, health centres and elderly care homes to warehouse and factory floors. The PTM is available with the GK indicator or AE 403 indicator, offering a range of features and functions. The GK indicator allows a numeric tare value to be entered, while the AE 403 is IP68 rated for dusty and wet environments."
Catalog No. NC2612396