Learn More
International Crystal PTFE IR Sample Cards, 50 per pack
Supplier:  International Crystal 00007363
Encompass_Preferred
PTFE IR Sample Cards, PTFE and polyethylene IR sample cards provide inexpensive disposable sample substrates for qualitative analysis, The 2 x 4 in. cards mount vertically in the universal slide mount in any spectrophotometer, 50/PK
Catalog No. NC9117494