Learn More
Revvity Health Sciences Inc Tag-lite T8-HaloTag (Zeocin) plasmid, 10 mg
Supplier:  Revvity Health Sciences Inc PT8HALOZEO
Encompass
Over the past few years, SNAP-Tag technology combined with TR-FRET has paved the road to the development of many non-radioactive, no-wash binding assays. The method is based on transfecting cells using plasmids encoding a SNAP-Tag and subsequently labeling them with Terbium. PT8HALOZEO is a backbone containing the gene for the HALO tag, a promoter a zeomycin resistance gene and MCS (Multiple Cloning Site).
Catalog No. 50-247-100