Learn More
Revvity Health Sciences Inc SNAP tag Beta2 adrenoR cloned plasmid, 1 Item
Supplier: Revvity Health Sciences Inc PSNAPBD2
Over the past few years, SNAP-Tag technology combined with TR-FRET has paved the way for the development of many non-radioactive, no-wash, binding assays. The method is based on transfecting cells using plasmids encoding a SNAP-Tag and subsequently labeling them with Terbium. Cisbio offers a large collection of such plasmids. All GPCR genes are cloned in an expression vector directly downstream from a CMV promoter, and are provided ready for protein expression and labeling. All information on this page pertains to the Tag-lite plasmid cloned with the Beta2 adreno receptor.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.