Learn More
Electron Microscopy Sciences PS25 Hardness Tester Calibration Vickers/Rockwell
Supplier: Electron Microscopy Sciences 6804905
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Reflected light calibration slide with diamond shapes and scales are used for for calibrating Vickers and Rockwell hardness testers. The PS25 has a series of diamond shapes of varying size and x-y scales, each clearly identified with its size. The glass disc has the image precision marked in vacuum deposited chrome and this is cemented into a stainless steel slide mount, making the calibration slide very durable.
The slide has a unique serial number indelibly marked on the slide mount and can be supplied with an Internationally traceable certificate of calibration. Supplied in a polished wooden box. Vickers/Rockwell.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.