Learn More
Fine Science Tools Providence-Hospital Hemostats, Stainless Steel, 14 cm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fine Science Tools 1301514
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.5mm/ Clamping Length: 15mm/ Alloy / Material: Stainless Steel/ Length: 14cm