Learn More
R&D Systems™ Proteome Profiler Human Protease Array Kit
The Proteome Profiler Human Protease Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
Supplier: R&D Systems™ ARY021B
This item is not returnable.
View return policy
Description
The Proteome Profiler Human Protease Array Kit contains 4 membranes - each spotted in duplicate with 35 different protease antibodies.Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
1 Kit | |
4 Array Membranes, 4-Well Multi-Dish, Array Buffers, Wash Buffer, Detection Antibody Cocktail, Streptavidin-HRP, Chemiluminescent Detection Reagents, Transparency Overlay Template, Detailed Protocol |