Learn More
Showa Denko America Inc PROTEIN LW-403 4D
Supplier:  Showa Denko America Inc F6989403
Encompass_Preferred
High performance type LW-400 series, superior efficiency versus LW-800 series, pore size specifically controlled for analyzing antibody type proteins, optimized for light scattering detection, high performance analysis of antibody drugs and various proteins, high lot-to-lot reproducibility, fulfills USP L20, L33, and L59 requirements, MW range (protein) = 10,000 to 700,000 Daltons, exclusion limit = 1,000,000 Daltons, dimensions 4.6 mm ID x 150 mm L
Catalog No. 50-161-3286