Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Protein LoBind™ Tubes
Protein LoBind™ Tubes are specifically designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
$202.50 - $246.00
Specifications
Closure Color | Gray |
---|---|
Color | Colorless |
Disposable | Single-use |
No. per Pack | 200 |
Packaging | Bag |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
E0030122216
|
Eppendorf™
0030122216 |
15 mL |
Each for $202.50
|
|
|||||
E0030122240
|
Eppendorf™
0030122240 |
50 mL |
Each for $246.00
|
|
|||||
Description
- Eppendorf™ LoBind™ material ensures optimized sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- Precise lid sealing to minimize evaporation
Specifications
Gray | |
Single-use | |
Bag | |
PCR Clean | |
Screw Cap | |
Yes |
Colorless | |
200 | |
Autoclavable | |
Conical | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title