Learn More
Showa Denko America Inc PROTEIN KW-802.5
Supplier: Showa Denko America Inc F6989000
SEC, GFC type silica-based packed columns for aqueous SEC (GFC) analysis, suitable for the analysis of proteins and enzymes, listed on USP L20, L33, and L59, MW range = 2,000 to 50,000 Daltons, exclusion limit = 60,000 Daltons, dimensions 8.0 mm ID x 300 mm L