Learn More
Labconco™ Protector™ Filtered Fume Hood Accessory, Roughing Prefilter
For use with Protector™ Echo™ and Airo™ Filtered Fume Hoods. Labconco™ Protector™ Filtered Fume Hood Accessory, Roughing Prefilter removes large particles to extend filter life.
Supplier: Labconco™ 9577402/EMD
Description
- Roughing prefilter
- Extends life of Neutrodine Filters (and HEPA Filters, if used)
- Located in light box
Warranty and Services
One year
Specifications
Roughing Prefilter | |
68.5 cm | |
1 in. | |
13 in. |
27 in. | |
Labconco Protector Echo and Airo Filtered Fume Hoods | |
2.54 cm | |
33 cm |