Learn More
Labconco™ Protector™ Filtered Fume Hood Accessories
Filters and accessories for use with Protector Echo and Airo Filtered Fume Hoods.
$119.00 - $1473.00
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
13-118-000
|
Labconco™
9577420 |
Neutrodine Unisorb Filter |
Each for $1,183.00
|
|
|||||
13-118-001
|
Labconco™
9577421 |
HEPA Filter |
Each for $1,040.00
|
|
|||||
13-118-002
|
Labconco™
9577422 |
Standard Prefilter |
Each for $119.00
|
|
|||||
13-118-003
|
Labconco™
9577423 |
Acid Prefilter |
Each for $164.00
|
|
|||||
13-118-004
|
Labconco™
9577424 |
Ammonia Prefilter |
Each for $182.00
|
|
|||||
13-118-005
|
Labconco™
9577433 |
Acid Sensor |
Each for $1,473.00
|
|
|||||
13-118-006
|
Labconco™
9577434 |
Formaldehyde Sensor |
Each for $1,473.00
|
|
|||||
Description
- These accessories are for use with Labconco Protector Echo and Airo Filtered Fume Hoods
- For all filters and prefilters: the number of filters/prefilters required depends on the width of the hood and if it is an initial order or a replacement order
Specifications
Protector Echo and Airo Filtered Fume Hoods |