Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Protecta™ Comfort Vest-Style Harness
Protecta™ Comfort Vest-Style Harness minimizes workday readjustment and is designed for comfort and convenience
$142.90 - $154.96
Specifications
Buckle Style | Pass-Thru; Parachute; Tongue Buckle |
---|---|
Capacity (English) | 420 lb. |
Capacity (Metric) | 190 kg |
Color | Black |
Material | Polyester |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-070-966
|
3M™
7012816690 |
Medium/Large |
Case for $142.90
|
|
|||||
19-070-967
|
3M™
7012816693 |
X-Large |
Case for $154.96
|
|
|||||
Description
- Fixed D-ring reduces the need for readjustment throughout the workday
- Specifically designed for added comfort
- Moisture-wicking, breathable back padding with soft edging for added comfort
- Modern design gives harness a fresh and sleek look
- ANSI-compliant auto-resetting lanyard keepers for fast and easy parking of snap hooks and carabiners
- Impact indicators make it easy to inspect the harness for prior damaging impact loads
- Protected labels for long-term identification and inspection
Specifications
Pass-Thru; Parachute; Tongue Buckle | |
190 kg | |
Polyester | |
Back | |
Polyester |
420 lb. | |
Black | |
Comfort Vest-Style Harness | |
3600 lb./ANSI |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title