Learn More
Ubpbio Proteasome Activity Fluorometric Assay Kit I
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ubpbio J4110
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Proteasome Activity Fluorometric Assay Kit I can be used for assaying the constitutive proteasomes' chymotrypsin-like activity in vitro using purified proteasome, cell lysates or tissue extracts. The proteasome cleaves Suc-LLVY-AMC, and the released AMC fluorescence can be monitored using a plate reader or fluorometer at the excitation/emission wavelength of 360nm/460nm, respectively.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.