Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ ProSpecT™ Giardia Microplate Assay
Rapidly detect Giardia specific antigen from human fecal samples.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | ProSpecT Giardia Microplate Assay |
Detectable Analytes | Giardia Cyst Antigen |
Sample Type | Fecal |
For Use With (Application) | Enzyme Immunoassay Detection |
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
R2458096
|
Thermo Scientific™
R2458096 |
ProSpecT Giardia Microplate Assay | 96 Tests/Kit | 2°C to 8°C |
Pack of 96 for $2,078.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Rapidly detect Giardia specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Giardia Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Giardia. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
Not all products are available for sale in all territories. Please inquire.
Specifications
2°C to 8°C | |
Giardia Cyst Antigen | |
Enzyme Immunoassay Detection |
ProSpecT Giardia Microplate Assay | |
Fecal | |
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title